Would you like to log in again?
EcoRI Adaptors, 150pmol, Promega
Supplier: Promega
C1291
PAC1291EA 69.07 USD InStock
PAC1291
EcoRI Adaptors, 150pmol, Promega
Nucleic Acid Reagents Primers and Probes
Attach EcoRI sticky ends to blunt-ended DNA
  • Streamline Cloning of Blunt-Ended DNA
  • Efficient addition of EcoRI restriction sites
  • Equimolar solution of two complementary oligonucleotides in water
  • Each oligonucleotide is 5’ phosphorylated
The EcoRI Adaptors consist of two complementary oligonucleotides: a 16mer and a 12mer phosphorylated at the 5'-end. The oligonucleotides are provided annealed in equimolar concentrations in water. The EcoRI Adaptors attach EcoRI sticky ends to blunt-ended DNA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty