Would you like to log in again?
FES Kinase Enzyme System, Promega
Supplier: Promega
V1981 V1982
PAV1981EA 612.3 USD InStock
PAV1981 PAV1982
FES Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human FES was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FES is a proto-oncogene that encodes a protein-tyrosine kinase distinct from c-Src, c-Abl and other nonreceptor tyrosine kinases.
  • Easily Screen and Profile FES Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human FES was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FES is a proto-oncogene that encodes a protein-tyrosine kinase distinct from c-Src, c-Abl and other nonreceptor tyrosine kinases. FES was originally identified as the cellular homolog of several transforming retroviral oncoproteins. FES plays a role in regulating cytoskeletal rearrangements and inside-out signalling that accompany receptor ligand, cell matrix and cell-cell interaction. Genetic analysis using a transgenic mouse model implicates FES in the regulation of inflammation and innate immunity. FES modulates the innate immune response of macrophages to LPS challenge, in part, by regulating the internalization and downregulation of the TLR4 receptor complex. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: FES, 10ug (Human, recombinant full-length). MW: ~125kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. FES NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2242/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty