Would you like to log in again?
Fetuin
Supplier: Promega
V4961
PAV4961EA 115.49 USD InStock
PAV4961
Fetuin
Proteins and Peptides
Fetuin is a glycoprotein with O-linked and N-linked glycosylation sites.
  • Glycoprotein with O- and N-Linked Glycosylation Sites
  • Deglycosylation substrate control for PNGase F, EndoH activity monitoring
  • Supplied at a concentration of 10mg/ml
Fetuin is provided as a deglycosylation substrate control. Fetuin is a glycoprotein with O-linked and N-linked glycosylation sites.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty