Would you like to log in again?
GoTaq® G2 Hot Start Polymerase, Promega
Supplier: Promega
GoTaq®
M7406 M7401 M7405 M7408
PAM7408EA 3941 USD InStock
PAM7408 PAM7405 PAM7406 PAM7401
GoTaq® G2 Hot Start Polymerase, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
GoTaq G2 Hot Start Polymerase is supplied with 5X Green GoTaq Flexi Buffer, 5X Colorless GoTaq Flexi Buffer and 25mM MgCl2.
  • High Performance GoTaq(R) G2 Enzyme with Convenient Room-Temperature Setup
  • Eliminate nonspecific amplification with a hot start enzyme
  • Detect lower-copy-number targets
  • Optimize reaction conditions with a Flexi Reaction Buffer and separate MgCl2
  • Supplied at a concentration of 5u/microl
For superior convenience and improved yield, sensitivity and specificity, choose GoTaq G2 Hot Start Polymerase, which is bound to a proprietary antibody that blocks activity. Activity is restored during initial denaturation, but is inhibited below 70 degrees C, allowing room-temperature reaction setup. GoTaq G2 Hot Start Polymerase is supplied with 5X Green GoTaq Flexi Buffer, 5X Colorless GoTaq Flexi Buffer and 25mM MgCl2. Hot-start PCR is advantageous for some amplification targets because it may eliminate or minimize primer-dimer and nonspecific products. In some cases, hot-start PCR may improve yields. GoTaq G2 Hot Start Polymerase exhibits 5' --> 3' exonuclease activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty