Would you like to log in again?
GoTaq Hot Start Colorless Master Mix, Promega
Supplier: Promega
GoTaq®
M5132 M5133
PAM5132EA 136.97 USD InStock
PAM5132 PAM5133
GoTaq Hot Start Colorless Master Mix, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
GoTaq Hot Start Master Mixes are all-in-one master mixes (just add template and primers) that contain the high-performance GoTaq DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two
  • Hot-Start GoTaq(R) DNA Polymerase in a Ready-to-Use Master Mix
  • All-in-one master mix: just add template and primers
  • Minimize nonspecific amplification and primer-dimers
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
GoTaq Hot Start Master Mixes are premixed, ready-to-use solutions containing GoTaq Hot Start Polymerase, magnesium, dNTPs and buffer. Reactions can be set up in less than a minute at room temperature; simply add your template, water and primers. GoTaq Hot Start Hot Start Polymerase contains the high-performance GoTaq DNA Polymerase bound to a proprietary antibody that blocks polymerase activity. The polymerase activity is restored during the initial denaturation step when the amplification reactions are heated at 94-95 degrees C for two minutes. This enables hot-start PCR, where polymerase activity is eliminated or minimized at temperatures below 70 degrees C. GoTaq Hot Start Polymerase exhibits 5' --> 3' exonuclease activity. The master mixes are available in green or colorless formats. The green master mix contain a compound that increases sample density so that samples sink easily into wells of an agarose gel as well as two dyes (yellow and blue) that separate to allow easy monitoring during electrophoresis. Use the green master mix for direct-to-gel analysis after amplification and the colorless master mix for amplifications where the dyes may interfere with post-amplification analysis such as fluorescence or absorbance testing. GoTaq Hot Start Master Mixes offer the specificity and sensitivity of an antibody-based hot-start polymerase in a convenient, easy-to-use, time-saving format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty