Would you like to log in again?
GoTaq Hot Start Polymerase, Promega
Supplier: Promega
GoTaq®
M5005 M5001 M5006 M5008
PAM5001EA 103.88 USD InStock
PAM5001 PAM5008 PAM5006 PAM5005
GoTaq Hot Start Polymerase, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
GoTaq Hot Start Polymerase contains the high-performance GoTaq DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two minutes.
  • Robust PCR with Convenient Room-Temperature Setup
  • Minimize nonspecific amplification and primer-dimers
  • Optimize reaction conditions with supplied Flexi Reaction Buffer and MgCl2 solution
  • Direct-to-gel convenience
  • Supplied at a concentration of 5u/µl
GoTaq Hot Start Polymerase contains the high-performance GoTaq DNA Polymerase bound to a proprietary antibody that blocks polymerase activity. The polymerase activity is restored during the initial denaturation step when the amplification reactions are heated at 94-95 degrees C for two minutes. This enables hot-start PCR, where polymerase activity is eliminated or minimized at temperatures below 70 degrees C. GoTaq Hot Start Polymerase exhibits 5' --> 3' exonuclease activity. The enzyme is supplied with a tube of 25mM MgCl2 to optimize the magnesium concentration in your reactions. It is also supplied with 5X Green GoTaq Flexi Buffer and 5X Colorless GoTaq Flexi Buffer. The buffers contain a compound that increases sample density so that samples sink easily into wells of an agarose gel. The green buffer also contains two dyes (yellow and blue) that separate to allow easy monitoring during electrophoresis. Use the green reaction buffer for direct-to-gel analysis after amplification and the colorless reaction buffer for amplifications where the dyes may interfere with post-amplification analysis such as fluorescence or absorbance testing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty