Would you like to log in again?
HaloCHIP System, 20 Reactions, Promega
Supplier: Promega
G9410
PAG9410EA 768.67 USD InStock
PAG9410
HaloCHIP System, 20 Reactions, Promega
Protein Purification Protein Purification Kits
The HaloCHIP System is designed for the covalent capture of intracellular protein:DNA complexes without the use of antibodies. An efficient alternative to the standard chromatin immunoprecipitation method.
  • Covalent Capture of Protein:DNA Complexes, No Antibodies Needed
  • No need to make or purchase qualified antibodies
  • Results in 24-48 hours with fewer steps, minimizing potential errors
The HaloCHIP System is a novel method designed for the covalent capture of intracellular protein:DNA complexes without the use of antibodies and offers an efficient and robust alternative to the standard chromatin immunoprecipitation (ChIP) method. Proteins of interest are expressed in cells as HaloTag fusion proteins, crosslinked to DNA with formaldehyde and then captured on HaloLink Resin, which forms a highly specific, covalent interaction with the HaloTag portion of the fusion protein. Stringent washing removes nonspecific proteins and DNA, and heating reverses the crosslinks between the DNA and the fusion protein and releases the captured DNA fragment, which subsequently can be purified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty