Would you like to log in again?
HaloTag NanoBRET 618 Ligand, 20 µl, Promega
Supplier: Promega
G9801
PAG9801EA 445.04 USD InStock
PAG9801
HaloTag NanoBRET 618 Ligand, 20 µl, Promega
Protein Linking and Labeling Systems
NanoBRET Transcriptional Protein Assays are sensitive, reproducible live-cell assays designed for monitoring or screening the interaction of proteins involved in transcriptional regulation.
  • Sensitive Live-Cell Assays for Monitoring Interaction of Proteins Involved in Transcription Regulation
  • Live-cell assay allows you to detect transcriptional protein interactions in real time using full-length proteins or fragments.
  • Monitor Changes: Reversible assay technology allows you to study both induction and inhibition of protein interactions.
NanoBRET Transcriptional Protein Assays are sensitive, reproducible live-cell assays designed for monitoring or screening the interaction of proteins involved in transcriptional regulation. Interactions between proteins are key events in the regulation of gene expression with protein homodimers and heterodimers interacting on DNA elements to regulate transcriptional events required for a variety of cellular responses. The NanoBRET cMyc/MAX Interaction Assay measures the specific interaction between the human cMyc and MAX transcription factors within their natural cellular context. The cMyc/MAX heterodimer regulates transcription related to cell proliferation, differentiation and apoptosis, making it an important candidate for drug targeting. NanoBRET assay technology is dependent upon energy transfer from a luminescent donor (NanoLuc luciferase) to a fluorescent acceptor (HaloTag NanoBRET 618 Ligand). NanoLuc luciferase and HaloTag protein are fused to the target proteins of interest and fusion proteins expressed at low cellular levels enabling monitoring and screening studies of protein interactions that reflect true cellular physiology. The NanoBRET assay is fully reversible, enabling studies of both induction and inhibition of protein interactions. For more details on NanoBRET Technology and a full list of available NanoBRET Transcriptional Protein Assays visit the NanoBRET Technology for Protein Interactions page.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty