Would you like to log in again?
HaloTEV Protease
Supplier: Promega
G6602 G6601
PAG6602EA 526.81 USD InStock
PAG6602 PAG6601
HaloTEV Protease
Proteins and Peptides
HaloTEV Protease (81kDa) is a fusion between the HaloTag protein and TEV protease, a highly specific proteolytic enzyme that cleaves at a specific TEV site, a specific seven-amino-acid sequence (ENLYFQ(G/S)).
  • Cleave and Recover Target Protein in a Single Step
  • Easy to covalently remove HaloTEV from your purified protein with HaloLink™ Resin
  • Proteins can be purified tag-free in a single step as the HaloLink™ Resin will bind both HaloTag(R) protein tag and the HaloTEV protease
  • Supplied at a concentration of 5u/microl
HaloTEV Protease (81kDa) is a fusion between the HaloTag protein and TEV protease, a highly specific proteolytic enzyme that cleaves at a specific TEV site, a specific seven-amino-acid sequence (ENLYFQ(G/S)). HaloTEV Protease allows covalent immobilization on HaloLink Resin and removal from a cleavage reaction in a single-step purification. The covalent capture of HaloTEV Protease improves purity of the final target protein and assures the improved recovery of the TEV protease.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty