Would you like to log in again?
HisLink Protein Purification Resin, Promega
Supplier: Promega
HisLink™
V8823 V8821
PAV8823EA 64.43 USD InStock
PAV8823 PAV8821
HisLink Protein Purification Resin, Promega
Protein Purification Protein Purification Resins
HisLink Protein Purification Resin is a macroporous silica resin modified to contain a high level of tetradentate-chelated nickel (>20mmol Ni/ml settled resin) for efficient capture and purification of bacterially expressed His- or HQ-tagged proteins.
  • Resin to Purify His- or HQ-Tagged Protein from Cleared Lysate
  • Purify polyhistidine- or HQ-tagged proteins from cleared lysate by standard column chromatography or from crude lysate in batch
  • Suitable for low- to medium-pressure applications such as FPLC
  • Large quantities of polyhistidine- or HQ-tagged protein can be purified per volume of resin used
HisLink Protein Purification Resin is a macroporous silica resin modified to contain a high level of tetradentate-chelated nickel (>20mmol Ni/ml settled resin) for efficient capture and purification of bacterially expressed polyhistidine- or HQ-tagged proteins. HisLink Resin may be used in either column or batch purification formats. Gravity feed of a cleared lysate over a HisLink column is sufficient for complete capture and efficient elution of polyhistidine- or HQ-tagged proteins; however, the resin also may be used with vacuum filtration devices (e.g., the Vac-Man Vacuum Manifold, Cat.# A7231) to allow simultaneous processing of multiple columns. HisLink Resin also is an excellent choice for affinity purification using low- to medium-pressure liquid chromatography systems such as fast performance liquid chromatography (FPLC). In batch format, HisLink Resin may be easily separated from the lysate without filtration, allowing processing of larger quantities of lysate and purification of protein from lysate that has not been cleared of insoluble cellular debris. Note: Resin product volumes represent total volume of a 50% resin slurry.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty