Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Trypsin
Supplier: Promega
V9013 V9012
PAV9012EA 322.34 USD InStock
PAV9012 PAV9013
Trypsin
Enzymes
Immobilized Trypsin provides a fast, convenient method for digesting a range of concentrations of purified protein or complex protein mixtures. Digested peptides are separated from the trypsin as they flow through the spin column into the collection tube.
  • Fast, Convenient Digestion of Purified Protein or Protein Mixtures
  • Digest a range of protein concentrations
  • Digestions completed in as little as 30 minutes
Immobilized Trypsin provides a fast and convenient method for digesting a range of concentrations of purified protein or complex protein mixtures. Digested peptides are easily separated from the Immobilized Trypsin as they flow through the spin column into the collection tube. Immobilized Trypsin is easily removed from the peptide solution because the trypsin does not pass though the column frit. Trypsin is a proteolytic enzyme, which cleaves at the carboxyl side of positively charged Lysine (Lys) and Arginine (Arg). When these amino acids are followed by the nonpolar Proline (Pro), the digestion of the site is not efficient. When Lys and Arg are followed by acids [Aspartic Acid (Asp) and Glutamic Acid (Glu)] the digestion is also not as efficient.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty