Would you like to log in again?
JM109 Competent Cells, >10(7)cfu/ug, Promega
Supplier: Promega
L1001
PAL1001EA 153.46 USD InStock
PAL1001
JM109 Competent Cells, >10(7)cfu/ug, Promega
Competent Cells
JM109 is a K strain that is recA- and endA- to minimize recombination and improve the quality of plasmid DNA. In addition, the cells carry the F´ episome, which allows blue/white screening.
  • Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
  • Cells are ready to use—no preparation time necessary
  • Transformation efficiencies guaranteed
  • No worries about undesirable recombination events (recA- mutation), and no nuclease carryover in miniprep DNA (endA- mutation)
JM109 is a K strain that is recA- and endA- to minimize recombination and improve the quality of plasmid DNA. In addition, the cells carry the F´ episome, which allows blue/white screening. JM109 Competent Cells are available for convenient transformation in two efficiencies: High Efficiency at greater than 10(8)cfu/ug and Subcloning Efficiency at greater than 10(7)cfu/ug. JM109 Competent Cells are prepared according to a modified procedure of Hanahan. JM109 Genotype: endA1, recA1, gyrA96, thi, hsdR17 (r(k)-, m(k)+), relA1, supE44, Delta(lac-proAB), [F' traD36, proAB, laqI(q)ZDeltaM15].
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty