Would you like to log in again?
K562 Genomic DNA, 80 µg, Promega
Supplier: Promega
E4931
PAE4931EA 351.29 USD InStock
PAE4931
K562 Genomic DNA, 80 µg, Promega
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
Purified from the human lymphoma cell line K562 by detergent lysis and proteinase K digestion. Supplied at a concentration of 400microg/ml in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).
  • Purified from the human lymphoma cell line K562 by detergent lysis and proteinase K digestion. Supplied at a concentration of 400microg/ml in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).
K562 Genomic DNA is purified from the human lymphoma cell line K562 by detergent lysis and proteinase K digestion. K562 Genomic DNA is supplied at a concentration of 400microg/ml in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty