Would you like to log in again?
Kemptide (PKA) Peptide Substrate
Supplier: Promega
V5601
PAV5601EA 79.79 USD InStock
PAV5601
Kemptide (PKA) Peptide Substrate
Proteins and Peptides
Kemptide is a peptide substrate for cAMP-Dependent Protein Kinase with the sequence Leu-Arg-Arg-Ala-Ser-Leu-Gly.
  • Specific Substrate for cAMP-Dependent Protein Kinases
  • Supplied ready for use in kinase reactions
Kemptide is a peptide substrate for cAMP-Dependent Protein Kinase with the sequence Leu-Arg-Arg-Ala-Ser-Leu-Gly.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty