Would you like to log in again?
LEV Elution Tubes, 50/pk, Promega
Supplier: Promega
AS6201
PAAS6201EA 76.5 USD InStock
PAAS6201
LEV Elution Tubes, 50/pk, Promega
Tubes Test Tubes
For use with the Maxwell(R) 16 and Maxwell(R) 16 forensic instruments in either SEV or LEV configuration.
  • Standard elution volume (SEV) is 200 to 400 µl
  • Low elution volume (LEV) is 30 to 100 µl
  • Temperatire range 15 to 30 °C
Plungers and elution tubes are only for the Maxwell(R) 16 instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty