Would you like to log in again?
LYN B Kinase Enzyme System, Promega
Supplier: Promega
V3712 V3711
PAV3711EA 623.02 USD InStock
PAV3711 PAV3712
LYN B Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human LYN B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LYN is a 56kDa tyrosine kinase that is similar to mouse T-lymphocyte-specific tyrosine kinase p56lck, v-yes, v-fgr and v-src.
  • Easily Screen and Profile LYN B Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human LYN B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LYN is a 56kDa tyrosine kinase that is similar to mouse T-lymphocyte-specific tyrosine kinase p56lck, v-yes, v-fgr and v-src and is expressed in a variety of tissues. LYN is expressed preferentially in B cells and can be coimmunoprecipitated with IgM, suggesting that LYN is physically associated with membrane-bound IgM and participates in antigen-mediated signal transduction. Crosslinking of membrane-bound IgM with antibody induces rapid increase in activities of LYN and LYN-associated phosphatidylinositol 3-kinase. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: LYN B, 10ug (Human, recombinant full-length). MW: ~85kDa. Substrate: SRC substrate (KVEKIGEGTYGVVYK-amide); derived from human p34cdc2 (amino acids 6-20). Other: Reaction Buffer, DTT, MnCl2. LYN B NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4067/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty