Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MagnaBot Spacer ¹/₈", 1 each, Promega
Supplier: Promega
V8581
PAV8581EA 51.11 USD InStock
PAV8581
MagnaBot Spacer ¹/₈", 1 each, Promega
Racks Magnetic Separation Racks
The spacers adjust the height of a 96-well plate placed onto the MagnaBot 96 Magnetic Separation Devices to optimize magnetization.
  • Adjust height of 96-well plate placed onto MagnaBot(R) 96 Magnetic Separation Devices
  • Optimize magnetization while using MagnaBot(R) 96 Magnetic Separation Devices
The MagnaBot Spacers are designed to optimize use of the MagnaBot 96 Magnetic Separation Device (Cat.# V8151) and the Deep Well MagnaBot 96 Magnetic Separation Device (Cat.# V3031). The spacers adjust the height of a 96-well plate placed onto the MagnaBot 96 Magnetic Separation Devices to optimize magnetization. Refer to specific robotic protocols and applications for recommended uses.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty