Would you like to log in again?
MagneGST Protein Purification System, Promega
Supplier: Promega
V8600 V8603
PAV8600EA 275.47 USD InStock
PAV8600 PAV8603
MagneGST Protein Purification System, Promega
Protein Purification Protein Purification Kits
MagneGST Protein Purification System provides a simple, rapid and reliable method for the purification of glutathione-S-transferase (GST) fusion proteins.
  • Simple Method for Glutathione-S-Transferase Fusion Protein Purification
  • One-step purification of multiple samples; no need for high-speed centrifugation after cell lysis
  • Use with 1-50ml of cell culture
  • Achieve high yields with little to no nonspecific background
The MagneGST Protein Purification System provides a simple, rapid and reliable method for the purification of glutathione-S-transferase (GST) fusion proteins. Immobilized glutathione paramagnetic particles (MagneGST Particles) are used to isolate GST-tagged protein directly from a crude or cleared lysate using either a manual or automated procedure and requires use of a magnetic stand. GST-tagged proteins can be purified on a small scale from 1ml of culture or on a large scale using more than 50ml of culture. Samples also can be processed using a robotic platform. MagneGST particles are supplied as a 25% slurry and have a binding capacity of 5-10mg of GST protein per 1ml of settled resin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty