Would you like to log in again?
MagneGST Pull-Down System, 80 reactions, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
V8870
PAV8870EA 928.12 USD InStock
PAV8870
MagneGST Pull-Down System, 80 reactions, Promega
Protein Purification Protein Purification Kits
MagneGST Pull-Down System detects protein interactions between GST-fusion proteins expressed in bacterial lysates and prey proteins expressed in the TNT Systems.
  • GST Pull-Down Assay for Protein:Protein Interaction Analysis
  • Analyze protein interactions between GST-fusion proteins from bacterial lysates and prey proteins expressed in TnT(R) Systems
The MagneGST Pull-Down System is designed for detection of protein interactions between GST-fusion proteins expressed in bacterial lysates and prey proteins expressed in the TNT Systems. Prey protein synthesized in the TNT Quick Coupled Transcription/Translation Reaction is captured using bait protein (GST-fusion protein) immobilized on MagneGST Particles. Nonspecifically bound proteins are then washed away, and the prey protein is analyzed. Prey proteins can be detected by incorporating radioactively labeled methionine in the TNT Quick reaction, followed by SDS-PAGE and autoradiography or by incorporating the supplied non-radioactive methionine in the TNT reaction and detecting by Western blotting with protein-specific antibodies.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty