Would you like to log in again?
BenchTop 100 bp DNA Ladder, 300 µl, Promega
Supplier: Promega
G8291
PAG8291EA 194.48 USD InStock
PAG8291
BenchTop 100 bp DNA Ladder, 300 µl, Promega
Electrophoresis Reagents Molecular Weight Markers
100-1,500bp in 100bp increments up to 1kb with an additional band at 1.5kb; 11 bands. Can be stored at room temperature. Contains dye marker, ready to load.
  • Eleven Fragments of 100-1,000bp in 100bp Increments Plus a Band at 1,500bp
  • Store at room temperature (22-25°C)
  • Premixed with loading buffer; ready to load onto agarose gels
  • Load 6microl/lane
The BenchTop DNA Markers offer the convenience of storage at room temperature (22-25°C) as well as the capability of direct loading onto agarose gels. The BenchTop DNA Markers are supplied in a stabilizing solution of 1X Blue/Orange Loading Dye, which circumvents any requirements for further manipulation. BenchTop 100bp DNA Ladder: Eleven fragments that range in size from 100bp to 1,000bp in 100bp increments with an additional band at 1,500bp. The 500bp fragment is present at increased intensity for easy identification. The ladder is dephosphorylated and ready for 5´ end-labeling with radioisotopes using T4 Polynucleotide Kinase, allowing visualization by autoradiography. Load 6microl/lane.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty