Would you like to log in again?
BenchTop PhiX174 DNA/HaeIII Markers, 250 µl, Promega
Supplier: Promega
G7511
PAG7511EA 224.49 USD InStock
PAG7511
BenchTop PhiX174 DNA/HaeIII Markers, 250 µl, Promega
Electrophoresis Reagents Molecular Weight Markers
72-1,353bp; 11 bands. Can be stored at room temperature. Contains dye marker, ready to load.
  • Eleven DNA Fragments of 72-1,353bp
  • Store at room temperature (22-25°C)
  • Premixed with loading buffer; ready to load onto agarose gels
  • Load 5microl/lane
The BenchTop DNA Markers offer the convenience of storage at room temperature (22-25°C) as well as the capability of direct loading onto agarose gels. The BenchTop DNA Markers are supplied in a stabilizing solution of 1X Blue/Orange Loading Dye, which circumvents any requirements for further manipulation. BenchTop ΦX174 DNA/HaeIII Markers: Eleven phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 72bp to 1,353bp. Load 5microl/lane.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty