Would you like to log in again?
p70S6K Kinase Enzyme System, Promega
Supplier: Promega
V2742 V2741
PAV2741EA 588.87 USD InStock
PAV2741 PAV2742
p70S6K Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human p70S6K was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. p70S6K is responsible for the phosphorylation of 40S ribosomal protein S6 and is ubiquitously expressed in human adult tissues.
  • Easily Screen and Profile p70S6K Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human p70S6K was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. p70S6K is responsible for the phosphorylation of 40S ribosomal protein S6 and is ubiquitously expressed in human adult tissues. p70S6K is activated by serum stimulation, and this activation is inhibited by wortmannin and rapamycin. p70S6K activity undergoes changes during the cell cycle and increases 20-fold in G1 cells released from G0. p70S6K activation requires sequential phosphorylations at proline-directed residues in the putative autoinhibitory pseudosubstrate domain, as well as threonine-389 a site phosphorylated by phosphoinositide-dependent kinase 1 (PDK-1). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: p70S6K, 10ug (Human, recombinant full-length). MW: ~76kDa. Substrate: S6K substrate (KRRRLASLR); derived from human 40S ribosomal protein S6 (amino acid 230-238). Other: Reaction Buffer, DTT. p70S6K NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/6198/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty