Would you like to log in again?
Pepsin
Supplier: Promega
V1959
PAV1959EA 30.82 USD InStock
PAV1959
Pepsin
Assays Enzyme Assays
Pepsin preferentially cleaves at the C-terminus of phenylalanine, leucine, tyrosine and tryptophan. Pepsin can be used alone or with other proteases for protein analysis by mass spectrometry and other applications. Activity is optimal at pH 1.0-3.0.
  • Cleaves at the C-Terminus of Phe, Leu, Tyr, Trp
  • Optimal activity at pH 1.0-3.0
  • Use for peptide mapping, protein identification, characterization of post-translational modifications
Pepsin preferentially cleaves at the C-terminus of phenylalanine, leucine, tyrosine and tryptophan. This protease can be used alone or in combination with other proteases for protein analysis by mass spectrometry and other applications. Pepsin activity is optimal at pH 1.0-3.0.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty