Would you like to log in again?
PhiX174 DNA/HaeIII Markers, 50 µg, Promega
Supplier: Promega
G1761
PAG1761EA 213 USD InStock
PAG1761
PhiX174 DNA/HaeIII Markers, 50 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Eleven phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 72bp to 1,353bp.
  • Eleven DNA Fragments of 72-1,353bp
  • Load 1microl/lane
  • Supplied at a concentration of 1µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
Conventional DNA Digest Markers are created by digesting either lambda DNA, PhiX174 replicative form DNA, or plasmids to completion with one or more restriction enzymes. The enzymes are heat-inactivated, and the DNA fragments are either phenol-extracted, then ethanol-precipitated or just ethanol-precipitated. The precipitated fragments are resuspended in storage buffer. The markers are not intended for quantitative analysis. Each marker is supplied with a tube of 6X Blue/Orange Loading Dye. PhiX174 DNA/HaeIII Markers: Eleven phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 72bp to 1,353bp. Recommended Loading: Load 1microl/lane.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty