Would you like to log in again?
PI3K (p120gamma)
Supplier: Promega
V1761
PAV1761EA 498.77 USD InStock
PAV1761
PI3K (p120gamma)
Enzymes
The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.
  • Purified PI3K Class I Lipid Kinases
  • Use with ADP-Glo™ Assay and PI3K Substrates in lipid kinase activity assays
Phosphatidylinositol (PI) and its phosphorylated derivates, collectively called phosphoinositides, are important second messengers that are critical as signaling molecules and for cellular membrane remodeling. The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty