Would you like to log in again?
PKC iota Kinase Enzyme System, Promega
Supplier: Promega
V3751 V3752
PAV3751EA 588.87 USD InStock
PAV3751 PAV3752
PKC iota Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human PKC iota was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C iota is a member of the protein kinase C family of serine-threonine kinases.
  • Easily Screen and Profile PKCι Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PKC iota was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C iota is a member of the protein kinase C family of serine-threonine kinases. The amino acid sequence of PKC iota showed greatest homology to PKC zeta, with 72% identity overall rising to 84% in the catalytic domain. PKC iota has been implicated in Ras signaling and is a critical downstream effector of oncogenic Ras in the colonic epithelium. Transgenic mice expressing constitutively active PKC iota in the colon are highly susceptible to carcinogen-induced colon carcinogenesis. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKC iota, 10ug (Human, recombinant full-length). MW: ~98kDa. Substrate: CREBtide (KRREILSRRPSYR); derived from human CREB1 isoform A (amino acid 109-121). Other: Reaction Buffer, DTT, Lipid solution. PKC iota NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5584/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty