Would you like to log in again?
Prime-a-Gene Labeling System, 30 reactions, Promega
Supplier: Promega
U1100
PAU1100EA 336.86 USD InStock
PAU1100
Prime-a-Gene Labeling System, 30 reactions, Promega
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
A complete set of complementary reagents, including Labeling 5X Buffer that contains random synthetic hexadeoxynucleotide primers for random-primed labeling of linear template DNA with radionucleotides.
  • Complete Kit for Random-Primed Labeling of Linear Template DNA, Excluding Radionucleotides
  • Ready-to-use reagents for random-primed labeling of linear DNA, including random hexamer primers (excludes radionucleotides)
  • Probes generated with high specific activities >1 x 109cpm/microg
The Prime-a-Gene Labeling System provides a complete set of complementary reagents, including Labeling 5X Buffer that contains random synthetic hexadeoxynucleotide primers for random-primed labeling of linear template DNA with radionucleotides. As little as 25ng of input DNA can be used to generate probes with specific activities >1 x 10(9)cpm/ug.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty