Would you like to log in again?
Protease Inhibitor Cocktail, 50X, Promega
Supplier: Promega
G6521
PAG6521EA 102.78 USD InStock
PAG6521
Protease Inhibitor Cocktail, 50X, Promega
Cell Culture Supplements and Additives
Protease Inhibitor Cocktail is used to prevent protein degradation after lysing cells. The product is a mixture of six different protease inhibitors with different target protease specificities.
  • Prevents Protein Degradation after Cell Lysis
  • Effective against many proteases
  • Best-in-class protease inhibition
  • Works with a wide array of fusion tags (e.g., Flag(R), His tag, GST) and capture technologies
Protease Inhibitor Cocktail is used to prevent protein degradation after lysing cells. The product is a mixture of six different protease inhibitors with different target protease specificities. The inhibitor cocktail is EDTA-free and provided as a powder, ready for reconstitution in 1ml of either 100% ethanol or DMSO to obtain a 50X working solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty