Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RNasin Ribonuclease Inhibitor, Promega

Supplier: Promega Corporation
N2111 N2115
PAN2115EA 692.71 USD InStock
PAN2115 PAN2111
RNasin Ribonuclease Inhibitor, Promega
Proteins and Peptides
Ribonuclease inhibitors with broad-spectrum RNase inhibitory properties. Recombinant and native available. Plus version protects RNA before, during and after heating.

  • Broad-Spectrum Inhibition of Common Eukaryotic RNases
  • Native, recombinant and oxidation-resistant forms available
  • Does not inhibit SP6, T7 or T3 RNA Polymerase; GoScript™ Reverse Transcriptase, AMV or M-MLV Reverse Transcriptase; or Taq DNA polymerase.
  • Use in many downstream assays: Inhibits a broad spectrum of eukaryotic RNases over a wide pH range (pH 5-8)
  • Maintains inhibitory activity over a wide temperature range

RNases are ubiquitous and can cause RNA degradation and compromise RNA integrity. Native and Recombinant RNasin Inhibitors are 50kDa proteins that inhibit RNase A family and human placental RNases by noncovalently binding to RNases in a 1:1 ratio. For downstream applications such as GoScript Reverse Transcriptase, AMV/M-MLV reverse transcriptases, SP6, T7/T3 RNA polymerase, and Taq DNA polymerases, Recombinant RNasin Inhibitor does not inhibit RNase T1, S1 nuclease, RNase from Aspergillus, RNase H, RNase ONE Ribonuclease and enzymes. RNasin Plus RNase Inhibitor is a recombinant mammalian RNase inhibitor that is expressed as a soluble protein in E. coli, allowing easy purification through a combination of ion exchange and hydrophobic interaction chromatography. The protein is capable of inhibiting eukaryotic RNases (e.g., RNase A and RNase B) similarly to human placental RNase inhibitor. RNasin Plus RNase Inhibitor is tested in RT-PCR and compatible with enzymes such as AMV, M-MLV and ImProm-II Reverse Transcriptases or Taq and Tfl DNA Polymerases. RNasin Plus RNase Inhibitor also is tested and compatible with quantitative, real-time RT-PCR in a TaqMan assay. RNasin Plus RNase Inhibitor offers increased resistance to oxidation over the human version of the protein. Two cysteines in the human protein have been identified as especially sensitive to oxidation and react by forming a disulfide bond that can block the active site of the inhibitor. RNasin Plus, through natural amino acid diversity, lacks the ability to form this site-blocking disulfide. In addition, the new protein has characteristics never before realized, including continued inhibition of RNases above 50°C. Heating solutions of RNasin Plus and RNase followed by cooling does not result in the reappearance of RNase activity—even when the solution is heated above the denaturation temperature of the RNasin Plus protein alone. This allows RNasin Plus to protect RNA species prior to, during and after heating, even at temperatures normally used during first-strand DNA synthesis in RT-PCR. Solutions heated up to 70°C for 15 minutes did not result in RNase reactivation.

Application: cDNA Synthesis, Transcription in vitro, Replication

Concentration: 20–40 units/µL

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty