Would you like to log in again?
Single-Stranded DNA, DNA Binding Protein, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
M3011
PAM3011EA 144.46 USD InStock
PAM3011
Single-Stranded DNA, DNA Binding Protein, Promega
Enzymes
Single-Stranded DNA Binding Protein binds with high affinity to single-stranded DNA but does not bind well to double-stranded DNA.
  • Cooperatively Binds ssDNA with High Affinity
  • Consists of four identical 18.9 kDa subunits
  • Does not bind well to double-stranded DNA
  • Supplied at a concentration of 1-5 µg/microl
E. coli Single-Stranded DNA Binding Protein (SSB) consists of four identical 18.9 kDa subunits. It binds with high affinity in a cooperative manner to single-stranded DNA but does not bind well to double-stranded DNA. It is involved in DNA replication and in recombination in vivo.
Applications: Replication, Recombination, Sequencing
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty