Would you like to log in again?
Star Printer Paper, 1 each, Promega
Supplier: Promega
E2161
PAE2161EA 111.39 USD InStock
PAE2161
Star Printer Paper, 1 each, Promega
Paper
These accessories are available for the now-discontinued 20/20 luminometer.
  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued
These accessories are only for the now-discontinued Turner BioSystems Luminometer Model TD-20/20. They are not for the Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax 20/20 Luminometer. Turner BioSystems and Promega have teamed up to develop the next-generation luminometers for single-tube and 96-well plate analysis. For more information, see the GloMax Luminometer catalog pages or contact Technical Support.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty