Would you like to log in again?
T4 DNA Ligase, Promega
Supplier: Promega
M1794
PAM1794EA 308.31 USD InStock
PAM1794
T4 DNA Ligase, Promega
Nucleic Acid Reagents Cloning Kits and Ligation
For ligation of DNA ends, mutagenesis and cDNA cloning
  • Joins Two DNA Strands with Cohesive or Blunt Ends
  • Use with 5´, 3´ or blunt-ended dsDNA (e.g., inserts and vectors)
  • Qualified for blue/white cloning, providing a higher level of quality control for enzymes used in cloning applications
  • Can catalyze the joining of RNA to either a DNA or RNA strand in a duplex molecule
T4 DNA Ligase catalyzes the joining of two strands of DNA between the 5'-phosphate and the 3'-hydroxyl groups of adjacent nucleotides in either a cohesive-ended or blunt-ended configuration. The enzyme has also been shown to catalyze the joining of RNA to either a DNA or RNA strand in a duplex molecule but will not join single-stranded nucleic acids.
Caution: Storage conditions –30...–10 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty