Would you like to log in again?
TNIK Kinase Enzyme System, Promega
Supplier: Promega
V6567 V4158
PAV4158EA 572.31 USD InStock
PAV4158 PAV6567
TNIK Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human TNIK (1-367) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TNIK or TRAF2 and NCK-interacting kinase are characterized by N-terminal kinase and C-terminal GCK domains that serve a regulatory function.
  • Easily Screen and Profile TNIK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human TNIK (1-367) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TNIK or TRAF2 and NCK-interacting kinase are characterized by an N-terminal kinase domain and a C-terminal GCK domain that serve a regulatory function (1). TNIK is mainly expression in brain, heart and spleen and is a specific effector of RAP2, which regulates actin cytoskeleton (2). TNIK is autophosphorylated in a manner dependent upon Lys54 in the ATP-binding pocket of its kinase domain and plays a main role in cytoskeleton regulation. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: TNIK, 10ug (Human, recombinant; amino acids 1-367). MW: ~67kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT, MnCl2. TNIK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/23043/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty