Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tween® 20 (Polysorbate)

Supplier: Promega Corporation
Synonyms: Polyoxyethylene sorbitan monolaurate

W3831
PAW3831EA 55 USD InStock
PAW3831
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Tween® 20 is a nonionic detergent used as a buffer component for immunoscreening in the ProtoBlot Systems.

Tween® 20 (100%) is used in immunoscreening to dissociate nonspecific interactions with an antibody probe.


Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty