Would you like to log in again?
Vac-Man 96 Vacuum Manifold, 1 each, Promega
Supplier: Promega
A2291
PAA2291EA 623.48 USD InStock
PAA2291
Vac-Man 96 Vacuum Manifold, 1 each, Promega
Manifolds
The Vac-Man 96 Vacuum Manifold is designed for manual use with the Promega SV 96 and SV 9600 Systems for isolating up to 96 samples simultaneously.
Eliminates the need to handle waste during nucleic acid purification
Adapts for use alone or with robotic nucleic acid purification platforms
Includes Vacuum Manifold Base, Manifold Collar and Manifold Bed
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty