Would you like to log in again?
Vac-Man Laboratory Vacuum Manifold, 20-sample capacity, 1 each, Promega
Supplier: Promega
A7231
PAA7231EA 210.21 USD InStock
PAA7231
Vac-Man Laboratory Vacuum Manifold, 20-sample capacity, 1 each, Promega
Manifolds
A vacuum manifold capable of processing 20 samples simultaneously. Supplied with 20 individual Luer-Lok Stopcocks, so each port is controlled individually.
Processes up to 20 samples simultaneously, while allowing individual control of each vacuum port
Eliminates the need to handle waste during nucleic acid purification
Includes Manifold, Stopcocks, Neoprene Stopper and Spinlock II Adapters
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty