Would you like to log in again?
Wizard Magnetic 96 DNA Plant System, Promega
Supplier: Promega
Wizard®
FF3760 FF3761
PAFF3760EA 289.51 USD InStock
PAFF3760 PAFF3761
Wizard Magnetic 96 DNA Plant System, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Wizard Magnetic 96 DNA Plant System is designed for manual or automated 96-well, high-throughput purification of DNA from plant leaf and seed tissue.
  • Automated Purification of DNA from Leaves and Seeds
  • Cuts purification time compared to CTAB extraction
  • Eliminates organic extractions
  • Validated for both leaf and seed tissue by PCR and RAPD analysis
The Wizard Magnetic 96 DNA Plant System is designed for manual or automated 96-well, high-throughput purification of DNA from plant leaf and seed tissue. The system has been validated with corn and tomato leaf, as well as with canola and sunflower seeds. The DNA purified from these samples can be used in PCR as well as more demanding applications such as RAPD analysis. Unlike column-based systems, the binding of nucleic acids to magnetic particles can occur in solution, enhancing contact with the wash buffer and increasing nucleic acid purity. Protocols are available for Beckman Coulter instruments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty