Would you like to log in again?
pmirGLO Dual-Luciferase miRNA Target Expression Vector, 20 µg, Promega
Supplier: Promega
E1330
PAE1330EA 514.26 USD InStock
PAE1330
pmirGLO Dual-Luciferase miRNA Target Expression Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
The pmirGLO Vector is designed to quantitatively evaluate microRNA (miRNA) activity by the insertion of miRNA target sites downstream or 3' of the firefly luciferase gene (luc2).
  • Evaluate microRNA Activity by Inserting miRNA Target Sites 3´ of the Firefly Luciferase Gene
  • Reporter activity correlates with miRNA activity
  • Optimized luc2 reporter gene provides highest expression while the Renilla luciferase gene provides normalization
  • The moderate-strength PGK promoter provides more biologically relevant analysis not possible with strong promoters
The pmirGLO Vector is designed to quantitatively evaluate microRNA (miRNA) activity by the insertion of miRNA target sites downstream or 3' of the firefly luciferase gene (luc2). Firefly luciferase is the primary reporter gene; reduced firefly luciferase expression indicates the binding of endogenous or introduced miRNAs to the cloned miRNA target sequence. This vector is based on Promega dual-luciferase technology, with luc2 used as the primary reporter to monitor mRNA regulation and Renilla luciferase (hRluc-neo) acting as a control reporter for normalization and selection.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty