Would you like to log in again?
pAdVAntage Vector, 20 µg, Promega
Supplier: Promega
E1711
PAE1711EA 197.59 USD InStock
PAE1711
pAdVAntage Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
Co-transfection of mammalian cells with the pAdVAntage Vector enhances transient protein expression in a variety of cell types by increasing translation initiation.
  • Enhances Protein Expression in Mammalian Cells When Cotransfected
  • Increases translation initiation in cells transiently transfected with protein expression constructs
  • Can be used in a variety of cell lines
Co-transfection of mammalian cells with the pAdVAntage Vector enhances transient protein expression in a variety of cell types by increasing translation initiation. Transfection of mammalian cells with an expression vector often results in suboptimal expression of the protein of interest. Double-stranded RNA (dsRNA) generated during transfection is thought to activate the dsRNA-activated inhibitor (DAI), one of several enzymes involved in the host cell's antiviral defense system. DAI phosphorylates the translation initiation factor eIF-2, halting translation and therefore protein production. However, DAI translation inhibition can be overcome with the adenoviral Virus Associated I RNA (VAI RNA) produced by RNA polymerase III following co-transfection with the pAdVAntage Vector. The VAI RNA binds to DAI, preventing its activation, thereby allowing translation and protein expression.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty