Would you like to log in again?
RT-PCR Kits, Agilent Technologies
Supplier: Agilent Technologies
600182 600182
ST600182EA 389.12 USD InStock
CAST600182 ST600182
RT-PCR Kits, Agilent Technologies
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Easy-A high-fidelity cloning enzymes combines the higher accuracy of a proofreading enzyme with the high cloning efficiency of Taq.
  • The only RT-PCR kit offering both TA/UA cloning and high fidelity
  • Produces PCR products ready for high-throughput TA/UA cloning with greater than 95% cloning efficiency
  • Preferentially adds a single 3'-A overhang
  • Accuracy level 6X greater than Taq
PCR products amplified with the Easy-A can be cloned directly, without performing additional steps typically required when amplifying with proofreading polymerases.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty