Would you like to log in again?
pGL4.83[hRlucP/Puro] Vector, 20 µg, Promega
Supplier: Promega
E7511
PAE7511EA 486.08 USD InStock
PAE7511
pGL4.83[hRlucP/Puro] Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
pGL4.82[hRluc/Puro], pGL4.83[hRlucP/Puro] and pGL4.84[hRlucCP/Puro] Vectors can accept putative promoter elements for investigation or control reporter vectors for dual-luciferase assays. Stably transfected cells can be selected using puromycin.
  • Cloning Vectors For Use as Control Reporter Vectors Offering Different Reporter Protein Stabilities
  • Designed for cloning a putative promoter element for investigating gene transcription control regions
  • Available with three varieties of engineered Renilla luciferase genes: hRluc, hRlucP or hRlucCP
  • Select for stably transfected cells using puromycin
Promoterless Renilla luciferase vectors are designed primarily to accept a putative promoter element for investigation of important regions controlling gene transcription. Alternatively, they may be used as promoterless control vectors in a dual-reporter system with a firefly luciferase vector serving as the experimental vector. The promoterless vectors are available with three varieties of engineered Renilla luciferase genes: hRluc, hRlucP or hRlucCP. The hRluc gene is engineered to remove most cryptic transcription factor binding sites and improve mammalian expression through codon optimization. The hRlucP and hRlucCP and RapidResponse genes are hRluc genes appended with degradation sequences to influence the cellular half-life of the hRluc gene. The RapidResponse genes respond more rapidly to stimuli but at the expense of signal intensity. The hRlucP gene responds more rapidly than hRluc2 with moderate signal intensity, and the hRlucCP responds more quickly with the lowest signal intensity. The promoterless pGL4.82[hRluc/Puro], pGL4.83[hRlucP/Puro] and pGL4.84[hRlucCP/Puro] Vectors are available with the selectable marker puromycin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty