Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

pH6HTC His6HaloTag T7 Vector, 20 µg, Promega

Supplier: Promega
G8031
PAG8031EA 313.21 USD InStock
PAG8031
pH6HTC His6HaloTag T7 Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
These multiple cloning site vectors are used for inducible expression of HaloTag-polyhistidine fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter.

  • Cloning Vectors with Dual Protein Fusion Tags for E. coli and Cell-Free Protein Expression
  • Choose from N- and C-terminal fusion options
  • Use either the polyhistidine tag or HaloTag(R) ORF for protein expression and analysis
  • Clone an protein coding region using PCR or restriction enzyme digestion

The following vectors are used for inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter. Expression levels depend highly on the nature of the protein, but in general the N-terminal HaloTag fusion protein can increase expression level, enhance refolding and boost solubility of the expressed protein. The MCS vectors contain a His6-HaloTag dual tag. The dual tag enables protein purification with the reusable Ni-resin while retaining the HaloTag covalent labeling properties. Multiple Cloning Site (MCS) Vectors: pH6HTN His6HaloTag T7 Vector (Cat.# G7971) is designed for protein expression with an N-terminal His6-HaloTag dual tag in E. coli and T7 cell-free expression systems. pH6HTC His6HaloTag T7 Vector (Cat.# G8031) is designed for protein expression with a C-terminal His6-HaloTag dual tag in E. coli and T7 cell-free expression systems.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty