Would you like to log in again?
pFC30A His6HaloTag T7 Flexi Vector, 20 µg, Promega
Supplier: Promega
G8321
PAG8321EA 322.02 USD InStock
PAG8321
pFC30A His6HaloTag T7 Flexi Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
These vectors are used for inducible expression of polyhistidine-HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter. Use the Flexi Cloning System vectors for C- or N-terminal fusions.
  • HaloTag(R)-Polyhistidine Dual Tag Bacterial Expression Vectors
  • Express proteins in E. coli T7 cell-free expression systems
  • Couple the protein solubility and labeling benefits of HaloTag(R) technology with the reusability and the throughput of Ni-affinity technology
  • Select N- or C-terminal tags and either Ampicillin or Kanamycin resistance
The following vectors are used for inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter. Expression levels depend highly on the nature of the protein, but in general the N-terminal HaloTag fusion protein can increase expression level, enhance refolding and boost solubility of the expressed protein. HaloTag vectors are supplied as Flexi System vectors. The Flexi Vector System is a simple, directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. Direct transfers can only occur between two N-terminal tagged vectors or from an N-terminal to a C-terminal vector. The Flexi system vectors contain a His6-HaloTag dual tag. The dual tag enables protein purification with the reusable Ni-resin while retaining the HaloTag covalent labeling properties. pFN29A/K His6HaloTag T7 Flexi Vectors (Cat.# G8261, G8331) are designed for protein expression with an N-terminal His6-HaloTag dual tag in E. coli T7 cell-free expression systems. pFC30A/K His6HaloTag T7 Flexi Vectors (Cat.# G8321, G8381) are designed for protein expression with a C-terminal His6-HaloTag dual tag in E. coli T7 cell-free expression systems. Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty