Would you like to log in again?
pFC27K HaloTag CMV-neo Flexi Vector, 20 µg, Promega
Supplier: Promega
G8431
PAG8431EA 322.02 USD InStock
PAG8431
pFC27K HaloTag CMV-neo Flexi Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
Vectors designed for expression of C-terminal-tagged HaloTag fusion proteins in mammalian cells. Once expressed, use the HaloTag fusion protein for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag Ligands.
  • HaloTag(R) C-Terminal Flexi(R) Vectors for Directional Cloning of Protein-Coding Sequences
  • Perform protein-coding region transfers between Flexi(R) Vectors without resequencing
  • Choose Ampicillin (A) or Kanamycin (K)
These vectors are designed for expression of C-terminal-tagged HaloTag fusion proteins in mammalian cells. Once expressed, the HaloTag fusion protein may be used for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag Ligands. In addition, the HaloTag fusion protein can be purified or pulled down as a complex with its protein partners. We offer two types of HaloTag fusion vectors to accommodate your cloning preferences: pHT Vector Series: Simple Multiple Cloning Site (MCS) plasmids for traditional cloning. pF Vector Series: Flexi Vector Cloning System---a directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert. Find your gene, precloned, and experimentally validated at: www.promega.com/products/pm/halotag-technology/kazusa-collection
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty