Would you like to log in again?
pTNT Vector, 20 µg, Promega
Supplier: Promega
L5610
PAL5610PK 266.39 USD InStock
PAL5610
pTNT Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
The pCMVTNT and pTNT Vectors are designed for convenient expression of cloned genes in vitro or in vivo. SP6 and T7 promoters allow expression from SP6- or T7-based coupled in vitro transcription/translation systems.
  • Convenient Expression of Cloned Genes In Vitro or In Vivo
  • Express cloned genes in vitro or in vivo from SP6- or T7-based coupled in vitro transcription/translation systems
The pCMVTNT and pTNT Vectors are designed for convenient expression of cloned genes in vitro or in vivo. SP6 and T7 promoters allow expression from SP6- or T7-based coupled in vitro transcription/translation systems. The presence of RNA phage promoters also allows highly efficient synthesis of RNA in vitro. Both vectors contain a 5´ beta-globin leader sequence and synthetic poly(A)30 tail, which have been shown to enhance expression of certain genes. For in vivo expression, the pCMVTNT Vector contains a CMV enhancer/promoter region, which allows strong constitutive expression in many cell types.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty