Would you like to log in again?
pSP64 Poly(A) Vector, 20 µg, Promega
Supplier: Promega
P1241
PAP1241EA 138.28 USD InStock
PAP1241
pSP64 Poly(A) Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
Used as standard cloning vector and for in vitro transcription from the SP6 promoter.
  • Standard Cloning Vector Used to Generate Poly(A)+ Transcripts In Vitro
  • In vitro transcription using the SP6 promoter next to the polylinker
  • Poly(A)+ transcripts generated by a stretch of 30 dA:dT residues inserted in the polylinker
  • Multiple cloning region provides a convenient selection of restriction sites for cloning
The pSP64 Poly(A) Vector can be used as a standard cloning vector and for in vitro transcription from the SP6 promoter. The pSP64 Poly(A) Vector also can be used to generate poly(A)+ transcripts in vitro. The vector has a stretch of 30 dA:dT residues inserted between the SacI and EcoRI sites. Therefore, when foreign DNA is cloned into any polylinker site other than EcoRI (HindIII, PstI, SalI, AccI, HincII, XbaI, BamHI, AvaI, SmaI or SacI), linearization of the recombinant plasmid with EcoRI allows the use of SP6 RNA polymerase in vitro to prepare RNA copies of the inserted sequences that contain a synthetic 3' poly(A) tail of 30 residues.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty