Would you like to log in again?
Alere iScreen® Saliva Alcohol Test Strips, Alere Toxicology
Supplier: Alere Toxicology
iScreen®
I-SA-25
75780-118BX 65.91 USD InStock
75780-118
Alere iScreen® Saliva Alcohol Test Strips, Alere Toxicology
Drug Tests
The iScreen® Saliva Alcohol Test Strip (Saliva) is a screening test used to semi-quantitatively measuring alcohol in human saliva. The test strip indicates relative Blood Alcohol Concentration (BAC) at 0.0%, 0.02%, 0.04%, 0.08%, and 0.30% cut-off levels.
The test is a firm plastic strip with a test pad attached at the tip. The test pad contains chemicals that are highly sensitive to alcohol. Alcohol in saliva reacts with the chemicals to produce a color change on the test pad. The color change depends on the amount of alcohol present. Results are read by comparing the color of the test pad with the color chart.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty