Would you like to log in again?
Cell Culture Dishes without Grid, Sarstedt
Supplier: Sarstedt
83.3902 83.3902.500
101093-427CS 262.88 USD InStock
101093-427 101093-377
Cell Culture Dishes without Grid, Sarstedt
Culture Dishes Cell Culture Dishes
TC-treated dishes allow convenient handling due to a raised ring around the circumference of the dish base. This enables an easy and secure grip of both the dish and the lid, even from a whole stack.
  • Polystyrene construction
  • Sterile
  • Pyrogen-free
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty