Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pig Pepsin (from Stomach mucosa)

Supplier: EMD Millipore
Calbiochem®
516360-500MG 516360-2.5GM 516360-500MG
80053-618EA 413.15 USD InStock
80053-618 80053-620 CA80053-620
Pig Pepsin (from Stomach mucosa)
Enzymes
Pepsin is isolated from procine stomach mucosa. Catalyzes the hydrolysis of aminoacyl-proline to an amino acid and proline. Inhibitors include aliphatic alcohols, pepstatin A and pH >6.0.

  • Lyophilized form
  • Store at —20 °C

Unit definition: One unit is defined as the amount of enzyme that will digest hemoglobin substrates to TCA-soluble products, measured as a change in absorbance at 280 nm of 0.001 per min at 37 °C, pH 2.0.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty