Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tween® 80 (Polysorbate) solution 10%, PROTEIN GRADE® detergent, Calbiochem®, Millipore®

Supplier: MilliporeSigma

655207-50ML
80057-686EA 164.9 USD InStock
80057-686
Tween® 80 (Polysorbate) solution 10%, PROTEIN GRADE® detergent, Calbiochem®, Millipore®
Tween® 80 (Polysorbate)
Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins. Supplied as a 10% (W/W) solution of PROTEIN GRADE® TWEEN® 80 detergent.

  • Aggregation number: 58
  • CMC: 12 μM
  • HLB number: 15
  • Conductivity: ≤50 µmhos

Packaging: Packaged under inert gas.


Storage Temperature: Refrigerator
MDL Number: MFCD00082107
CAS Number: 9005-65-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty